Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Z163473

Dricap® dehydrator cartridge

weight 1.0 g (desiccant), L × W 1/2 in. × 19/32 in.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41100000
Product Z163473 is not currently sold in your country. Contact Technical Service

manufacturer/tradename

Multisorb 02-00033AG35

L × W

1/2 in. × 19/32 in.

weight

1.0 g (desiccant)

Looking for similar products? Visit Product Comparison Guide

General description

DriCap is a safe, non-toxic and non-dusting sorbent canister that provides exceptional moisture adsorbing capacity in a small space. With a number of sorbent types available, DriCap can protect a variety of packaged products from the effects of moisture including caking, mildew, fermentation, potency loss and decreased shelf life.

They have a slower adsorption rate for longer handling times. Contains super-activated indicating silica gel. Some typical applications for use are in pharmaceuticals, medical diagnostics kits, electronic equipment and optical instruments.

Example: One DriCap Desiccant Canister - with a nominal weight of 0.75 g, a nominal diameter of 0.34 in. (8.73 mm) and a nominal length of 0.875 in. (22.2 mm) - when sealed in a volume of 10 in.3 (163.87 cm3) at 77°F (25°C) and 70% RH, will reduce that atmosphere to less than 30% RH within 12 hours.

Legal Information

Dricap is a registered trademark of Multisorb Technologies, Inc.

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service