Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Z163392

Sigma-Aldrich

Lab-desiccator cartridge

cartridge diam. 114 mm

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
24141516
NACRES:
NB.43

description

for 160 mm diam. desiccator

manufacturer/tradename

Bel-Art F42045-0160

cartridge diam.

114 mm

Looking for similar products? Visit Product Comparison Guide

General description

The lab desiccator cartridge is conveniently sized to fit in any desiccator. It can be regenerated repeatedly, without losing drying performance. With a perforated, see-through Mylar® top and a long-wearing, permeable felt bag, all the seams are also heat sealed. NatraSorb® TI indicating silica gel change color to pink when saturated.

To regenerate the saturated gel cartridges, simply place in a vented 50°C (300°F) oven for 3 or more hours (depending on size) until blue color returns. Packaged in moisture-barrier pouch.

Legal Information

Mylar is a registered trademark of Dupont Teijin Films
NatraSorb is a registered trademark of Multisorb Technologies, Inc.

pictograms

Health hazardEnvironment

signalword

Danger

Hazard Classifications

Aquatic Chronic 2 - Carc. 1B - Repr. 1B

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service