Key Documents
Z162426
Diamond-needle files
set contains half-round, round, square files
About This Item
UNSPSC Code:
41100000
NACRES:
NB.35
General description
Set of five containg, half-round, round, and square files. All are 14 cm L with 2 in. diamond surface bonded to faces. Use in deburring, smoothing, and finishing ultrahard materials such as glass. Comes in vinyl pouch.
Choose from one of the most recent versions:
Certificates of Analysis (COA)