Key Documents
Z160482
Sigma-Aldrich
Safety drum faucet
brass body, pkg of 1 ea
About This Item
UNSPSC Code:
41100000
NACRES:
NB.51
material
brass body
packaging
pkg of 1 ea
manufacturer/tradename
Justrite 8902
General description
Self-closing, FM approved. 3/4 in. NPT thread, rigid, brass.
Choose from one of the most recent versions:
Certificates of Analysis (COA)