Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

Z159832

Cochranes atom centers for Orbit molecular models

iodine, univalent, dark green

Synonym(s):

chemistry model sets, molecular model sets

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
60104506
NACRES:
NB.17

manufacturer/tradename

Cochranes 0223

color

dark green

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service