Key Documents
Z156914
Justrite® nonmetallic wide-spout disposal can
capacity 5 gal
About This Item
UNSPSC Code:
41100000
manufacturer/tradename
Justrite 14755
H × O.D.
20 in. × 12 in.
capacity
5 gal
General description
Handles hazardous spent liquids. Resilient red HDPE resists physical abuse and is impervious to most solvents, fuels, and corrosive chemicals. Self-closing cap, 35/8 in. pouring spout, pressure relief valve, and SS flame arrester. Withstands 50 psig without rupture or structural damage. FM approved. Cadmium-plated fittings.
Legal Information
Justrite is a registered trademark of Justrite Manufacturing Co.
Choose from one of the most recent versions:
Certificates of Analysis (COA)