Key Documents
Z156868
Justrite® oval laboratory can
capacity 1 gal
About This Item
UNSPSC Code:
41100000
NACRES:
NB.35
material
red high-density polyethylene
stainless steel fitting
manufacturer/tradename
Justrite 14160
W × D × H
4 5/8 in. × 7 5/8 in. × 12 3/4 in.
capacity
1 gal
General description
Tough, seamless, resilient red HDPE body safety cans resist physical abuse and are impervious to most solvents, fuels, and corrosive chemicals. Self-closing pressure-relief cap and SS flame arrester. Withstands 50 psig without rupture or structural damage. FM approved.
Legal Information
Justrite is a registered trademark of Justrite Manufacturing Co.
Choose from one of the most recent versions:
Certificates of Analysis (COA)