Key Documents
Z156825
Sigma-Aldrich
Justrite® Type 1 safety can
capacity 5 gal
About This Item
UNSPSC Code:
41100000
NACRES:
NB.35
material
terneplated steel
manufacturer/tradename
Justrite 10801
H × O.D.
17 in. × 11 1/2 in.
capacity
5 gal
General description
For the safe transport and storage of gasoline and other flammable liquids. All cans have a double brass, wire-mesh flame arrester and a self-closing spout for automatic pressure-relief. Not for use with corrosive solvents or solvents with water. Red enamel finish. FM approved and UL listed. Meets OSHA requirements.
Legal Information
Justrite is a registered trademark of Justrite Manufacturing Co.
Choose from one of the most recent versions:
Certificates of Analysis (COA)