Key Documents
Z156760
Sigma-Aldrich
Adjustable-capacity drum pump
Fits 5, 15, 30, or 55 gal containers, PP/PE
About This Item
UNSPSC Code:
41100000
NACRES:
NB.51
feature
Fits 5, 15, 30, or 55 gal containers
General description
Adjustable capacity, dispenses liquids at a rate of 2, 4, 6, or 8 oz/downstroke depending upon which adapter is in place. Uses: most acids and alkalis.
Choose from one of the most recent versions:
Certificates of Analysis (COA)