Key Documents
Z155977
Stainless steel drying trays
rolled bead, L × W × H 17 in. × 11.6 in. × 0.75 in.
About This Item
UNSPSC Code:
41104510
NACRES:
NB.43
manufacturer/tradename
Polarware 17F
L × W × H
17 in. × 11.6 in. × 0.75 in.
General description
Stainless steel drying trays have a low profile, and are lighter weight than glass. We carry a wide range of sizes. The unbreakable stainless steel material has rapid cooling and heating for your samples.
Choose from one of the most recent versions:
Certificates of Analysis (COA)