Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Z153788

Mininert® Valve, screw thread

screw-cap valve, cap, 13 mm-425

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122001

feature

cap 13 mm-425

Looking for similar products? Visit Product Comparison Guide

General description

These valves are very easy to use.
1. Push the green button to open the valve
2. Insert the needle through the septum and take sample
3. Remove the needle and push to close
Benefits include:
  • PTFE construction for inertness. PTFE is useful when working when acids and organic solvents.
  • Good sealing ability for containing most volalite liquids and gases at low pressures
  • Mininert valves can be used at temperatures up to 105 °F.
  • 0.036 in. (0.9 mm) opening accepts up to 20-gauge needle

Silicone septa, red

Application

Mininert valves are leak-tight closures for vials and other containers. They are available in both crimp and screw thread versions. Only PTFE and glass are in contact with the contents of the container. They are the ideal closure for the following applications:
  • calibration standards
  • air- or moisture-sensitive fluids
  • derivatizating reagents
  • volatile chemicals

Other Notes

The maximum temperature that Mininert Valves can be used at is 105° F. When Mininert Valves are used at high temperatures, returning them to room temperature may cause the valve to leak slightly.

Legal Information

Mininert is a registered trademark of Valco Instruments Co. Inc.

replaced by


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service