Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Z150614

Thermometer for MEL-TEMP® apparatus

glass (mercury-filled)

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
UNSPSC Code:
41112203

material

glass (mercury-filled)

manufacturer/tradename

Barnstead International 1004

Looking for similar products? Visit Product Comparison Guide

Legal Information

MEL-TEMP is a registered trademark of Barnstead/Thermolyne Corp.

signalword

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Aquatic Acute 1 - Aquatic Chronic 1 - Repr. 1B - STOT RE 1

Storage Class

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service