Key Documents
Z150460
Keck® Joint Clip
standard taper, Set of 5 each for Joint sizes 10, 14, 19, 24, 29
About This Item
UNSPSC Code:
41122413
NACRES:
NB.35
General description
Color coded, Delrin® plastic joint clips will not scratch glassware or corrode. Withstand temperatures up to 302 °F (150 °C). Resistant to concentrated alkalis and diluted acids; not resistant to halogen gases or concentrated acids.
Comes in a plastic storage box.
Set of 5 each for Joint Size 10 (green), 14 (yellow), 19 (blue), 24 (green), and 29 (red).
Legal Information
Delrin is a registered trademark of E. I. du Pont de Nemours and Company
Keck is a registered trademark of Adcuram Lab Technologies GmbH
Choose from one of the most recent versions:
Certificates of Analysis (COA)