Z150312
Aldrich® septum-inlet adapter with side-arm
joint: ST/NS 24/40
All Photos(1)
About This Item
UNSPSC Code:
41121815
joint
joint: ST/NS24/40
inlet diam.
14 mm
General description
Requires septum Z564702 or Z564699.
Septum-inlets require septum Z565695 or Z565679 except where noted.
Linkage
See the Threaded Ground Glass Tutorial to see how threaded joints work.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)