Key Documents
Z150274
Sigma-Aldrich
Kilo-Lab® transfer lines
stainless steel valve
About This Item
UNSPSC Code:
41100000
NACRES:
NB.43
material
stainless steel valve
General description
Convenient fully assembled lines for removing liquid from Kilo-Lab cylinders. Line consists of a brass CGA 510 fitting connected to a 48in. length of metal braided PTFE tubing connected to a metal ball valve with metal needle connector. "T" is placed in-line and is equipped with a needle valve and 6mm I.D. septum inlet for nitrogen flush of transfer line.
Legal Information
Kilo-Lab is a registered trademark of Merck KGaA, Darmstadt, Germany
Choose from one of the most recent versions:
Certificates of Analysis (COA)