Key Documents
Z147346
Supelco
ACE PTFE tubing connector
About This Item
UNSPSC Code:
23151817
NACRES:
SB.52
manufacturer/tradename
Ace Glass 585307
technique(s)
LPLC: suitable
General description
Connects 1/2 in. I.D. tubing to #15 Ace-Thred. Fits Ace chromatography columns.
Choose from one of the most recent versions:
Certificates of Analysis (COA)