Key Documents
Z146781
Swagelok® tube connector
brass, for tubing o.d., 1/4 in.
About This Item
UNSPSC Code:
41121800
NACRES:
NB.51
Pricing and availability is not currently available.
material
brass
feature
for tubing o.d. 1/4 in.
General description
1/4 in. NPTF inlet to tube outlet.
Legal Information
Swagelok is a registered trademark of Swagelok Company
Choose from one of the most recent versions:
Certificates of Analysis (COA)