Key Documents
Z144061
Pyrex® pennyhead stopper
size 19
Synonym(s):
glass stopper, glass stoppers, stoppers, Corning® 7660
About This Item
UNSPSC Code:
41122500
material
glass
size
19
manufacturer/tradename
Corning
L
17 mm , ground-zone
General description
Ground zone length indicates its greatest diameter (mm).
Legal Information
Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)