Key Documents
Z137510
Sigma-Aldrich
Aldrich® Abderhalden drying apparatus replacement parts
Desiccant flask
About This Item
UNSPSC Code:
41121800
material
borosilicate glass
joint
joint SJ40/35 F
stopcock size
2 mm
General description
The desiccant flask is a replacement part for the Aldrich Abderhalden drying apparatus, Z219673.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)