Key Documents
Z137146
Supelco
Ace chromatography column
I.D. × L 25 mm × 51 cm
About This Item
UNSPSC Code:
41121804
NACRES:
SB.52
manufacturer/tradename
Ace Glass 590245
technique(s)
LPLC: suitable
I.D. × L
25 mm × 51 cm
suitability
suitable for chromatography
General description
Has #15 Ace-Thred at top and 2 mm bore, solid-PTFE stopcock-plug (tapered 1:5) at bottom. Supplied with nylon and FETFE® O-ring for connecting 12.5 to 14 mm O.D. tube to column, and with tubing connector.
Note:
When using tubing connector, O-ring supplied with bushing is NOT necessary. Fritted disc has 70 to 100 μm porosity, sealed into bottom of column.
Legal Information
FETFE is a registered trademark of Ace Glass, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)