Key Documents
Z136484
Pipette fillers
blue natural rubber bulb type
About This Item
UNSPSC Code:
41121500
NACRES:
NB.01
material
blue natural rubber bulb
General description
Valved, choice of natural or silicone rubber bulb. Allows safe manual control of filling and discharging of liquids by finger pressure.
Choose from one of the most recent versions:
Certificates of Analysis (COA)