Key Documents
Z135909
Sigma-Aldrich
Write-on label tape
W 1 in.
About This Item
UNSPSC Code:
55121611
material
paper
white
packaging
pkg of 3 rolls
manufacturer/tradename
Bel-Art F13483-0100
W
1 in.
General description
For applying labels to surfaces of any contour or material (glass, metal, plastic). Requires no special stylus: just pencil, ballpoint pen, or solvent ink marker. Resists moisture and most solvents, and can be used at temperatures of -73 to 121 °C. Will withstand repeated autoclaving and remain legible under most lab use. May be readily applied and peeled off without leaving sticky residue. 40 yd roll.
Legal Information
Write-on is a trademark of SP Scienceware, a division of SP
Choose from one of the most recent versions:
Certificates of Analysis (COA)