Key Documents
Z135690
Scienceware® Polystop dropper bottle
capacity 60 mL
Synonym(s):
bottle dropper, dropper bottles
About This Item
UNSPSC Code:
41121800
NACRES:
NB.17
material
colorless bottle
glass bottle
round bottle
joint
joint ST/NS neck
packaging
pack of 1 ea
manufacturer/tradename
Bel-Art F11604-0060
capacity
60 mL
General description
Use dropper without removing stopper. Clear glass bottle. Dropper with PP stopper and soft-vinyl bulb. Ground, tapered neck on bottle fits stopper. Can be disassembled to clean parts separately.
Legal Information
Polystop is a trademark of SP Scienceware, a division of SP
Scienceware is a registered trademark of SP Scienceware, a division of SP
Choose from one of the most recent versions:
Certificates of Analysis (COA)