Key Documents
Z135658
Burette clamp
double
Synonym(s):
burette clamps, clamp lab, lab clamp
About This Item
UNSPSC Code:
41121800
NACRES:
NB.43
General description
Noncorroding HDPE. Concealed steel spring and rubber pads prevent slipping. Center clamp tightens to fit standard support rod; 1/2 and 3/8 in. bushings included.
Choose from one of the most recent versions:
Certificates of Analysis (COA)