Z134244
Whatman® PTFE membrane filters
PTFE membrane circles, (TE 36), 0.45 μm, 25 mm, 50/pk
Synonym(s):
Whatman filter, Whatman filters, ptfe filter
About This Item
packaging
pack of 50
manufacturer/tradename
Whatman 10411305
Whatman Article No. 28415314 (US reference)
diam.
25.0 mm
pore size
≤0.45 μm pore size
suitability
suitable for (applications involving aggressive organic solvents, strong acids, and alkalis)
Looking for similar products? Visit Product Comparison Guide
General description
High chemical resistance and mechanical strength
One of the major applications for the PTFE membrane is the clarification of corrosives, solvents, and aggressive fluids. This includes the important requirement in HPLC analysis for sample filtration where any solid particles can cause permanent damage to the column. The 0.5 µm pore size is normally used. Air and gas sterilization make use of the hydrophobic characteristics of PTFE membrane filters and their ability to stop aqueous aerosols. Pore sizes of 0.2 µm and 0.5 µm are typically used. Sterile* venting of vacuum manifolds, fermentation vessels, and filtrate tanks and containers utilize PTFE 0.2 µm membranes.
Application
- Clarification of corrosives, solvents, and aggressive fluids
- HPLC
Other Notes
The legacy GE Whatman products that use the SIAL "Z" product numbering system will be discontinued and directly replaced with a product number using a "WHA" prefix plus the GE product code. For reference, the legacy "Z" product number is identified as a synonym on the "WHA" product number listing.
Legal Information
replaced by
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service