Z134139
Whatman® weighing boats and paper
B-2 Sheets, 6 x 6 inch, 500/pk
Synonym(s):
weigh paper, weighing paper
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41100000
material
paper
square
feature
grade B-2
packaging
pack of 500
manufacturer/tradename
Whatman 10347673
Whatman Article No. 28414664 (US reference)
size
6 in. × 6 in.
Looking for similar products? Visit Product Comparison Guide
Other Notes
This product has been replaced by WHA10347673-ALDRICH | Whatman weighing boats and paper, B-2 Sheets, 6 x 6 inch, 500/pk
Global SIAL customers (excluding Japan) can now order any product from the Cytiva Life Sciences consumables portfolio (prepack only) using GE′s product numbering system.
The legacy GE Whatman products that use the SIAL "Z" product numbering system will be discontinued and directly replaced with a product number using a "WHA" prefix plus the GE product code. For reference, the legacy "Z" product number is identified as a synonym on the "WHA" product number listing.
The legacy GE Whatman products that use the SIAL "Z" product numbering system will be discontinued and directly replaced with a product number using a "WHA" prefix plus the GE product code. For reference, the legacy "Z" product number is identified as a synonym on the "WHA" product number listing.
Legal Information
Whatman is a registered trademark of Cytiva
replaced by
Product No.
Description
Pricing
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service