Key Documents
Z126314
Supelco
Conversion kit for chromatography sprayer
About This Item
UNSPSC Code:
41115711
grade
for chromatography
manufacturer/tradename
Ace Glass 8464999
suitability
suitable for chromatography
Other Notes
Converts Z12,630-6 to 50ml capacity. Includes 50ml glass tube and hexfoot stand.
Choose from one of the most recent versions:
Certificates of Analysis (COA)