Key Documents
Z124990
Sigma-Aldrich
Syringe PP/PE without needle
syringe set, 25 Luer slip tip syringes: 5 each of 3, 5, 10, 20, and 50 mL, sterile
Synonym(s):
polypropylene syringes
About This Item
UNSPSC Code:
42142600
NACRES:
NB.03
sterility
sterile
General description
These polypropylene syringes with Luer tips, may be used with any size needle. The rigid polyethylene plungers of these disposable syringes are slightly oversized, creating a tight seal against the walls of the semi-flexible PP barrel, as well as providing an exceptionally smooth draw. No contaminating lubricants added.
Syringes are individually packaged (sterile) except for bulk packed products; these are supplied in plastic bags (non-sterile).
Order needles separately.
Choose from one of the most recent versions:
Certificates of Analysis (COA)