Key Documents
Z124486
Miniature Inert Valve
4 port, distribution plug, Hamilton (HV) plug valve
About This Item
UNSPSC Code:
41122001
NACRES:
NB.03
parameter
−10-100 °C temperature (at 500 psi)
General description
For building inert liquid or gas flow systems that will handle small volumes of ultrapure and corrosive fluids. Has universal housing with 1/4 × 28 in. female threaded ports that accept a wide variety of interchangeable fittings. Requires less than 2 in.3 of space.
Choose from one of the most recent versions:
Certificates of Analysis (COA)