material
cellulose matrix
glass support
feature
binder Proprietary
fluorescent indicator: no
manufacturer/tradename
Sigma-Aldrich
technique(s)
thin layer chromatography (TLC): suitable
L × W
20 cm × 20 cm
layer thickness
250 μm
particle size
20 μm
Looking for similar products? Visit Product Comparison Guide
General description
- Silica gel - for weakly to strongly polar separations, pore size 60 Å
- High purity silica gel - acid washed for separation of aflatoxins
- Cellulose - for partition chromatography, (SigmaCell Type 100 Cellulose)
- Cellulose PEI - for separation of weak anions (amino acids, peptides) PEI plates may turn slightly yellow, affecting separation. Pre-develop plates in distilled water to minimize yellow color.
- Chiral silica - for separation of optically active isomers by ligand exchange, pore size 60Å
Application
Other Notes
105716
Legal Information
replaced by
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service