Z122807
Sigma-Aldrich® TLC Plates
silica, highly purified matrix, L × W 20 cm × 20 cm
About This Item
Recommended Products
material
polyester support
silica, highly purified matrix
feature
binder Gypsum/polymeric
fluorescent indicator
manufacturer/tradename
Sigma-Aldrich
technique(s)
thin layer chromatography (TLC): suitable
L × W
20 cm × 20 cm
layer thickness
250 μm
plate L × W
20 cm × 20 cm
particle size
17 μm
Looking for similar products? Visit Product Comparison Guide
General description
- Silica gel - for weakly to strongly polar separations, pore size 60 Å
- High purity silica gel - acid washed for separation of aflatoxins
- Cellulose - for partition chromatography, (SigmaCell Type 100 Cellulose)
- Cellulose PEI - for separation of weak anions (amino acids, peptides) PEI plates may turn slightly yellow, affecting separation. Pre-develop plates in distilled water to minimize yellow color.
- Chiral silica - for separation of optically active isomers by ligand exchange, pore size 60Å
Application
Other Notes
100390
Legal Information
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service