Z118753
Spinbar® magnetic stirring fleas
yellow, diam. 1.5 mm, L 8 mm
About This Item
material
PTFE-coated
cylindrical PTFE-coated bar
feature
ALNICO V coated magnet
packaging
pack of 3 ea
manufacturer/tradename
Bel-Art F37121-0002
L
8 mm
diam.
1.5 mm
color
yellow
General description
- Spinbar® magnetic stirring fleas are designed for use in tubes, vials, and gradient markers.
- Ideal for mixing of small volume samples in environmental testing and life science applications.
- Alnico magnet encapsulated in durable and inert FDA grade PTFE (polytetrafluoroethylene) coating.
- Broad chemical resistance for use in a wide variety of applications.
- Autoclavable.
- PTFE-coated
- ALNICO V coated magnet
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)