Key Documents
Z118672
Air-driven stirrer
Light usage, parameter 200-10000 rpm
Synonym(s):
air driven stirrers, air mixer, air stirrer
About This Item
UNSPSC Code:
41103801
NACRES:
NB.72
packaging
pack of 1 ea
manufacturer/tradename
MODEL A
parameter
200-10000 rpm
support rod diam. × L
9/16 in. × 10 in.
General description
  • Explosion-proof, stall-proof, does not overheat
  • Exhaust muffler
  • Brass needle valve with 1/4 in. NPTM outlet controls air intake
  • Mount on standard clamp holders and vertical rods (order separately)
Includes: a screw collet and 3/8 in. diam. × 12 in. L SS stirrer shaft with propeller. A 3-jaw geared chuck may be ordered separately.
Choose from one of the most recent versions:
Certificates of Analysis (COA)