Key Documents
Z118206
Sigma-Aldrich
Angled septum-inlet adapter
brass, inlet I.D. 6 mm × 6 mm
About This Item
UNSPSC Code:
41121815
NACRES:
NB.43
material
brass
inlet I.D.
6 mm × 6 mm
General description
1/4 in. NPTM × top inlet × side inlet for septa.
Linkage
See the Threaded Ground Glass Tutorial to see how threaded joints work.
Other Notes
6 mm I.D. inlets require septa Z565709 or Z565717.
Choose from one of the most recent versions:
Certificates of Analysis (COA)