Key Documents
Z117439
LDPE laboratory tubing
I.D. × O.D. 1/8 in. × 1/4 in.
About This Item
UNSPSC Code:
41100000
I.D. × O.D.
1/8 in. × 1/4 in.
wall thickness
1/16 in.
General description
Good chemical resistance, translucent, and rigid. Temp. range: -103 to 120°F.
Choose from one of the most recent versions:
Certificates of Analysis (COA)