Key Documents
Z115754
Sigma-Aldrich
Aldrich® short condenser
joint: ST/NS 24/40, jacket L 175 mm
Select a Size
1 EA
$865.00
$865.00
Available to ship onApril 29, 2025Details
Select a Size
Change View
1 EA
$865.00
About This Item
UNSPSC Code:
41121800
NACRES:
NB.43
$865.00
Available to ship onApril 29, 2025Details
joint
joint: ST/NS24/40
jacket L
175 mm
Related Categories
General description
Useful in distilling solids. May be used for conventional distillations as a condenser or a jacketed column.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)