Key Documents
Z114251
Sigma-Aldrich
Plier-type decapper
O.D. 13 mm
About This Item
UNSPSC Code:
41122001
NACRES:
NB.11
O.D.
13 mm
General description
Removes aluminum seals from serum bottles and vials.
Choose from one of the most recent versions:
Certificates of Analysis (COA)