Key Documents
Z113522
Sigma-Aldrich
Electrode connector adapter
US Standard meter to BNC electrode
About This Item
UNSPSC Code:
41115603
NACRES:
NB.79
General description
If the type of connector on your pH meter is not compatible with your electrode, choose an appropriate adapter.
Choose from one of the most recent versions:
Certificates of Analysis (COA)