Key Documents
Z112003
Sigma-Aldrich
Demountable liquid-cell kit
About This Item
UNSPSC Code:
12352200
NACRES:
SB.32
General description
Useful for studying mulls, films, solutions, and viscous liquids. Demountable windows make re-polishing and replacement easy. Pathlengths can be changed simply by using different spacers. Kit includes one cell holder, one PTFE O-ring and two gaskets, one aluminum plate, two PTFE stoppers, one space compensator for use with Irtran-2, and one each of six different pathlength spacers (0.015, 0.025, 0.05, 0.1, 0.2, 0.5 mm). Order 32 mm windows separately. Liquid cells require a drilled front window and an undrilled back window. Order replacement parts separately.
Replacement parts:
PTFE stopper, Z111996
PTFE gasket, Z112011
PTFE O-ring, Z176613
Choose from one of the most recent versions:
Certificates of Analysis (COA)