Key Documents
Z111813
Sigma-Aldrich
Hydrion® Insta-Check® 0-13 pH test paper
L × W 15 ft × 7/32 in.
About This Item
UNSPSC Code:
41113034
NACRES:
NB.79
feature
non-bleeding: no
refill: no
packaging
pkg of 2 rolls
manufacturer/tradename
Hydrion
L × W
15 ft × 7/32 in.
detection method
colorimetric
General description
A wide range, direct-reading, general-purpose pH paper with color chart of 14 colors, pH range 0 to 13. Test paper is supplied in convenient dispenser packages each with its own color chart.
Legal Information
Hydrion is a registered trademark of Micro Essential Laboratory, Inc.
Insta-Check is a registered trademark of Micro Essential Laboratory, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)