Key Documents
Z111384
Sigma-Aldrich
Wig-L-Bug® grinder/mixer, analog
AC/DC input 115 V AC
About This Item
UNSPSC Code:
12352200
NACRES:
SB.32
feature
CE compliant (UL Approved)
availability
not available in EU
General description
Provides a fast, simple and inexpensive method for mixing, grinding and collecting samples which are consistent from batch to batch. Has 60 second timer, and comes with a vial adapter for 1/2 in. diam. × 1 in. long vials.
Legal Information
Wig-L-Bug is a registered trademark of Dentsply Rinn
Choose from one of the most recent versions:
Certificates of Analysis (COA)