Z105961
Wrap-it ties
L 5.6 in.
Synonym(s):
tie wrap, wrap tie
All Photos(1)
About This Item
UNSPSC Code:
41121800
NACRES:
NB.35
parameter
30 lb tensile strength
L
5.6 in.
General description
Self-locking, one-piece nylon construction. Excellent for securing rubber septa to bottles and flask joints, attachment of tubing to hose barbs or glassware, and for closing plastic bags or drum liners. Use with Wrap-it tie installing tool.
Tear-off version is blue in color and is easily removed from septum with a quick pulling motion.
Tear-off version is blue in color and is easily removed from septum with a quick pulling motion.
Choose from one of the most recent versions:
Certificates of Analysis (COA)