Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Z105880

Cyanoacrylate adhesives

Low-viscosity

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
UNSPSC Code:
31201600
NACRES:
NB.35

description

Ethyl Cyanoacrylate

viscosity

30-50 cP

Looking for similar products? Visit Product Comparison Guide

General description

  • Fast setting
  • Chemically resistant
  • Supplied in 1 oz bottles
  • High strength
  • Bond similar and dissimilar materials
  • Temp. range: -80 to 180 °C

Physical properties

Shear strength: 2200psi

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

185.0 °F - closed cup

flash_point_c

85 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service