Z104868
PTFE sleeve
joint: ST/NS 10/30
Synonym(s):
ptfe sleeves
All Photos(1)
About This Item
UNSPSC Code:
41100000
Recommended Products
joint
joint: ST/NS10/30
packaging
pkg of 6 ea
manufacturer/tradename
Bel-Art
General description
Extremely thin, accurately tapered, slip-on tubes for ground-glass joints prevent seizing and galling. Chemically inert and used without lubricants or adhesives, thus precluding contamination. Temperature range: −100 to 250°C.
Choose from one of the most recent versions:
Certificates of Analysis (COA)