Key Documents
Z104000
Fieser molecular model research kit
color-coded plastic and aluminum parts
About This Item
UNSPSC Code:
60104506
NACRES:
NB.17
Product Z104000 is not currently sold in your country. Contact Technical Service
General description
Low-cost molecular models designed by Professor Louis F. Fieser. The models consist of sturdy, color-coded plastic and aluminum parts that snap together to form bonds. They are easily assembled and handled, and may be disassembled for repeated use. Fieser models are highly effective for assessing conformational and steric effects, and geometrical relationships. Additional atoms and bonds are available separately. Each kit consists of:
  • 30 tetrahedral carbon atoms (black)
  • 6 pairs of double-bonded carbon atoms (black)
  • 5 oxygen (or sulfur) atoms (red)
  • 2 trivalent nitrogen atoms (blue)
  • 2 sulfur atoms (yellow)
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Visit the Document Library
L.F. Fieser
Journal of Chemical Education, 42, 408-408 (1965)
L.F. Fieser
Journal of Chemical Education, 40, 62-62 (1963)
L.F. Fieser
Journal of Chemical Education, 40, 457-457 (1963)