X1105
o-Xylylenebis(triphenylphosphonium bromide)
97%
Synonym(s):
(o-Phenylenedimethylene)bis[triphenylphosphonium bromide], 1,2-Bis(triphenylphosphoniomethyl)benzene dibromide, NSC 665614, NSC 78589
About This Item
assay
97%
reaction suitability
reaction type: C-C Bond Formation
mp
>300 °C (lit.)
SMILES string
[Br-].[Br-].C(c1ccccc1C[P+](c2ccccc2)(c3ccccc3)c4ccccc4)[P+](c5ccccc5)(c6ccccc6)c7ccccc7
InChI
1S/C44H38P2.2BrH/c1-7-23-39(24-8-1)45(40-25-9-2-10-26-40,41-27-11-3-12-28-41)35-37-21-19-20-22-38(37)36-46(42-29-13-4-14-30-42,43-31-15-5-16-32-43)44-33-17-6-18-34-44;;/h1-34H,35-36H2;2*1H/q+2;;/p-2
InChI key
MXCBXCYGWQAOSN-UHFFFAOYSA-L
Application
- Synthesis of [6.8]3cyclacene and polyunsaturated [23] and [24]metacyclophanes
- Selective inclusion of equatorial isomers of cyclohexane-polyols in phosphonium salt hosts
- Photoactive platinum(II) DNA intercalator synthesis
- Preparation of (pheynlenedivinylene)bis[pyrrole] derivatives
- The reaction with sodium hexamethyldisilazide to form acyclic, ring, and cage AS,C compounds
- Synthesis of styryl substituted annelated furan derivatives
signalword
Warning
hcodes
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
target_organs
Respiratory system
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service