WHA113502
Whatman® Nuclepore™ Track-Etched Membranes
L × W 8 in. × 10 in., pore size 0.03 μm
Synonym(s):
Z748846
About This Item
Recommended Products
sterility
non-sterile
packaging
pk of 25
manufacturer/tradename
Whatman 113502
Whatman Article No. 28417760 (US reference)
L × W
8 in. × 10 in.
pore size
0.03 μm
Looking for similar products? Visit Product Comparison Guide
Application
- Epifluorescence microscopy
- Environmental analysis
- Cell biologyEPA testing
- Fuel testing
- Bioassays
- Parasitology
- Air analysis
- Water microbiology
Other Notes
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service