Key Documents
WHA10537173
Whatman® protein saver card accessories
903 Dry rack without velcro, pack of 10 ea
Synonym(s):
Z699535, dry rack, protein saver card
About This Item
UNSPSC Code:
41111700
packaging
pack of 10 ea
manufacturer/tradename
Whatman 10537173
Whatman Article No. 28416980 (US reference)
General description
Whatman Dry Rak is designed to accommodate multiple collection forms at one time, properly air-drying the blood specimens in a suspended horizontal position. The Dry Rak is easily assembled and can be affixed to a wall or counter top with optional Velcro stickers.
Other Notes
Field of Use : For internal research use only. Products are not intended for diagnostic use or resale.
Legal Information
Whatman is a registered trademark of Cytiva
Choose from one of the most recent versions:
Certificates of Analysis (COA)