Key Documents
WHA10499001
Whatman® membrane marking pen
Synonym(s):
Whatman filter, Whatman membrane, Z699500, membrane filter, membrane pen
About This Item
UNSPSC Code:
41111700
NACRES:
NB.79
packaging
pkg of 10 ea
manufacturer/tradename
Whatman 10499001
Whatman Article No. 28416536 (US reference)
General description
For marking nitrocellulose and nylon transfers
The membrane marking pen is a high-xylene, felt-tipped marker that permanently writes on nitrocellulose and nylon membranes used in standard transfer procedures.
This indispensable tool ensures easy identification and orientation of gel transfers, colony and plaque lifts, and Western blots, even when the membrane is damp. It is compatible with hybridization and incubation buffers. An ideal marker for keeping records of transfers.
Other Notes
Field of Use : For internal research use only. Products are not intended for diagnostic use or resale.
Legal Information
Whatman is a registered trademark of Cytiva
Choose from one of the most recent versions:
Certificates of Analysis (COA)