WHA10463898
Whatman® Roby automated filter 25 mm
filter validation kit
Synonym(s):
Z751731
About This Item
Recommended Products
material
polypropylene housing
various membrane
sterility
non-sterile
packaging
pk of 1
manufacturer/tradename
Whatman 10463898
Whatman Article No. 28415905 (US reference)
diam.
25 mm
pore size
0.45-1.0 μm
Looking for similar products? Visit Product Comparison Guide
General description
The filter housing is made from mechanically stable polypropylene. The external geometry of the filter housing ensures simple and smooth filter transport from the storage turntable to the filtration site and easy filter changing.
Features
Optimized for Sotax, Caliper, Zymark® and Varian® tablet testers
Mechanically stable polypropylene
Benefits
Easy filter changing
Ensures simple and smooth filter transport
Applications
Fine filtration of samples in the automatic tablet dissolution test
Method development with the Roby 25 Filter Validation Kit
Features and Benefits
- The kit includes one tube of each membrane type
- Syringe filters included: nylon, regenerated cellulose, regenerated cellulose with GF92, glass fiber GF55, and glass fiber GF92
- At-a-glance information about the properties of each filter
- Filter selection aid to select the ideal filter of the six available options
- Includes filter validation protocol
- Optimized for SOTAX, Caliper (Zymark®), Varian®, and Erweka tablet tester systems
Components
Filter Validation Kit includes: Roby 25/GF92; Roby 25/GF55; Roby 25/RC; Roby 25/RC-GF92; Roby 25 NL; Roby 25 NL-GF92. (6 tubes of 25 pieces each
Suitability
Other Notes
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service